Ethyl hydrogen malonate 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Mono-Ethyl malonate, 3-Ethoxy-3-oxopropanoic acid

39990-0050 39990-0250 39990-2500
TS39990-0050EA 47 USD
TS39990-0050 TS39990-0250 TS39990-2500
Ethyl hydrogen malonate 95%
Ethyl hydrogen malonate
Formula: C₅H₈O₄
MW: 132.115 g/mol
Boiling Pt: 106.5 °C (3 mmHg)
Density: 1.119 g/cm³
Storage Temperature: Refrigerator
MDL Number: MFCD00020490
CAS Number: 1071-46-1

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Titration with NaOH ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR