Water HPLC

Supplier: Thermo Scientific Chemicals

Synonyms: Hydrogen oxide

389390010 389390025
TS389390010EA 63 USD
TS389390010 TS389390025
Water HPLC
Water
Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100
Melting Pt: 0
Density: 1 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
Merck Index: 13,10098

Order Now

Specification Test Results

Appearance Clear colorless Liquid
UV at 254 nm A: ≤0.004
Residue after evaporation ≤0.0003 %
Conductivity ≤1 µS/cm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR