Sodium octadecyl sulphate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Sodium Stearyl sulphate, n-Octadecyl sulphate, sodium salt, Sodium n-octadecylsulphate, Octadecyl sulphuric acid salt

38489-0010 38489-0050 38489-0250
TS38489-0010EA 78 USD
TS38489-0010 TS38489-0050 TS38489-0250
Sodium octadecyl sulphate 98%
Sodium octadecyl sulphate
Formula: C₁₈H₃₇NaO₄S
MW: 372.55 g/mol
Melting Pt: 211…213 °C
Storage Temperature: Ambient
MDL Number: MFCD00007469
CAS Number: 1120-04-3

Order Now

Specification Test Results

Appearance White to light yellow to light beige Powder
Infrared spectrum Conforms
Loss on drying ≤7 %
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR