Ethyl-4-oxocyclohexanecarboxylate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ethyl 4-oxocyclohexanecarboxylate, 4-(Ethoxycarbonyl)-1-cyclohexanone

37991-0010 37991-0050 37991-2500
TS37991-0010EA 29 USD
TS37991-0010 TS37991-0050 TS37991-2500
Ethyl-4-oxocyclohexanecarboxylate 97%
Ethyl-4-oxocyclohexanecarboxylate
Formula: C₉H₁₄O₃
MW: 170.21 g/mol
Boiling Pt: 150…152 °C (40 mmHg)
Density: 1.076 g/cm³
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00013285
CAS Number: 17159-79-4

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4580 to 1.4610 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR