3-Bromopropionaldehyde dimethylacetal 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Bromo-1,1-dimethoxypropane

37626-0050
TS37626-0050EA 186 USD
TS37626-0050
3-Bromopropionaldehyde dimethylacetal 95%
3-Bromopropionaldehyde dimethylacetal
Formula: BrH₂CCH₂CH(OCH₃)₂
MW: 183.05 g/mol
Boiling Pt: 58…60 °C (14 mmHg)
Flash Pt: 46 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00013245
CAS Number: 36255-44-4
EINECS: 252-936-6
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4460 to 1.4500 (20 °C, 589 nm)
GC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR