Tricine 99+% for biochemistry

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-((1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl)amino)acetic acid, Tricine buffer, N-(Tris(hydroxymethyl)methyl)glycine

32783-0010
TS32783-0010EA 532 USD
TS32783-0010
Tricine 99+% for biochemistry
Tricine
Formula: (HOCH₂)₃CNHCH₂CO₂H
MW: 179.17 g/mol
Melting Pt: 180…184 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00004277
CAS Number: 5704-04-1
EINECS: 227-193-6
Merck Index: 13,09724

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Water ≤1 % (K.F.)
UV at 280 nm A: ≤0.02
pH 4.6 to 5.6 (1 % in water at 25°C)
Titration with HClO4 ≥99.0 %
Lead (Pb) ≤5 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR