Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) 99.85% for molecular biology DNase-, RNase-, protease-free

Supplier: Thermo Scientific Chemicals

Synonyms: 2-Amino-2-(hydroxymethyl)propane-1,3-diol(tris), Tromethamine, Tris(hydroxymethyl)methylamine, 2-Amino-2-(hydroxymethyl)propane-1,3-diol, 2-Amino-2-(hydroxymethyl)-1,3-propanediol, Trizma(R) base, TRIS base, THAM

32736-0010 32736-0050
TS32736-0010EA 151 USD
TS32736-0010 TS32736-0050
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) 99.85% for molecular biology DNase-, RNase-, protease-free
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol)
Formula: NH₂C(CH₂OH)₃
MW: 121.14 g/mol
Boiling Pt: 357
Melting Pt: >168.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00004679
CAS Number: 77-86-1
EINECS: 201-064-4
Merck Index: 13,09842
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11a03d1ab6b2c11b9a2c83f0a73fc4264cec0110&vid=5ce890cdf298a59a7900a07fee1c857a5e5e61e0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals
Infrared spectrum Conforms
Water ≤1 %
Solubility (10 % in water) Clear and free of haze
UV at 260 nm A: ≤0.03
pH 10.0 to 11.7 (5 % at 25°C)
Titration with HClO4 ≥99.80 %
Dnase,Rnase,Protease activity None detected
Arsenic (As) ≤5 ppm
Copper (Cu) ≤1 ppm
Iron (Fe) ≤1 ppm
Lead (Pb) ≤1 ppm
Magnesium (Mg) ≤1 ppm
Insoluble matter ≤0.0005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR