Zinc acetate dihydrate 98+% ACS

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Acetic acid zinc salt dihydrate, Zinc (II) acetate dihydrate, Zinc diacetate dihydrate

AC31721-0025 AC31721-5000
TS31721-0025EA 170 USD
TS31721-0025 TS31721-5000
Zinc acetate dihydrate 98+% ACS
Zinc acetate dihydrate
Formula: (H₃CCOO)₂Zn·2H₂O
MW: 219.51 g/mol
Melting Pt: 237
Density: 1.84 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00066961
CAS Number: 5970-45-6
EINECS: 209-170-2
UN: 3077
ADR: 9 III
Merck Index: 12,10256

Order Now

Specification Test Results

Appearance White Crystals
Titration Complexometric 98.0 to 101.0 %
pH 6.0 to 7.0 (5 % at 25°C)
Chloride (Cl) ≤5 ppm
Sulfate (SO4) ≤0.005 %
Antimony (Sb) ≤2 ppm
Calcium (Ca) ≤0.005 %
Iron (Fe) ≤5 ppm
Lead (Pb) ≤0.002 %
Magnesium (Mg) ≤0.005 %
Potassium (K) ≤0.01 %
Sodium (Na) ≤0.05 %
Insoluble matter ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR