L-Serinamide hydrochloride 98%

Supplier: Thermo Scientific Chemicals

Synonyms: (S)-2-Amino-3-hydroxypropanamide hydrochloride, H-Ser-NH₂·HCl

303160010 303162500
TS303160010EA 50 USD
TS303160010 TS303162500
L-Serinamide hydrochloride 98%
L-Serinamide hydrochloride
Formula: C₃H₉ClN₂O₂
MW: 140.567 g/mol
Melting Pt: 184…190 °C
Storage Temperature: Ambient
MDL Number: MFCD00067560
CAS Number: 65414-74-6
New Product

Order Now

Specification Test Results

Appearance White to off-white Powder or crystals
Infrared spectrum Conforms
Melting point 184 to 190 °C
Titration Argentometric ≥97.5 %
Specific optical rotation +13.5° to +15.5° (20 °C, 589 nm) (c=1, H2O)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR