Tetraethylammonium tetrafluoroborate 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Tetrylammonium tetrafluoroborate

AC22161-0250 AC22161-1000 AC22161-5000
TS22161-0250EA 108 USD
TS22161-0250 TS22161-1000 TS22161-5000
Tetraethylammonium tetrafluoroborate 99%
Tetraethylammonium tetrafluoroborate
Formula: C₈H₂₀BF₄N
MW: 217.06 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00011827
CAS Number: 429-06-1
EINECS: 207-055-1

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals
Infrared spectrum Conforms
Solubility (4 % in acetonitrile) Clear to almost clear colorless
Elemental Analysis ≥98.5 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR