Mercury(II) chloride 99.5%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Mercuric chloride, Mercury Chloride, Mercury dichloride

AC20143-0010 AC20143-5000
TS20143-0010EA 337 USD
TS20143-0010 TS20143-0250 TS20143-1000 TS20143-5000
Mercury(II) chloride 99.5%
Mercury(II) chloride
Formula: HgCl₂
MW: 271.5 g/mol
Boiling Pt: 302 °C (760 mmHg)
Melting Pt: 277 °C
Storage Temperature: Ambient
MDL Number: MFCD00011041
CAS Number: 7487-94-7
EINECS: 231-299-8
UN: 1624
ADR: 6.1 II
Merck Index: 13,05901

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals (May contain black particles)
Titration Complexometric ≥99.4 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR