18-Crown-6 (1,4,7,10,13,16-hexaoxacyclooctadecane) 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,4,7,10,13,16-Hexaoxacyclooctadecane, 18-Crown-6

18156-0050 18156-0250 18156-1000 18156-5000
TS18156-0050EA 48 USD
TS18156-0050 TS18156-0250 TS18156-1000 TS18156-5000
18-Crown-6 (1,4,7,10,13,16-hexaoxacyclooctadecane) 99%
18-Crown-6 (1,4,7,10,13,16-hexaoxacyclooctadecane)
Formula: C₁₂H₂₄O₆
MW: 264.32 g/mol
Boiling Pt: 396
Melting Pt: 36…40 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00005113
CAS Number: 17455-13-9
EINECS: 241-473-5
Merck Index: 13,02630

Order Now

Specification Test Results

Appearance Clear colorless to white Crystals or crystalline mass or liquid
Infrared spectrum Conforms
Melting point 36 to 40 °C
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR