Sodium cyanate 85%, pure

Supplier: Thermo Scientific Chemicals
Warning

AC18056-2500
TS18056-2500EA 58 USD
TS18056-2500
Sodium cyanate 85%, pure
Sodium cyanate
Formula: CNNaO
MW: 65.01 g/mol
Melting Pt: 550 °C
Storage Temperature: Ambient
MDL Number: MFCD00011121
CAS Number: 917-61-3
EINECS: 213-030-6
Merck Index: 12,08749

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Titration Argentometric ≥80 %
Loss on drying ≤3 % (5 g, 105°C, 2 hrs)
Sodium carbonate (Na2CO3) ≤15 %
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR