Silver cyanide 99% for analysis

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Argentous cyanide

AC18023-0100
TS18023-0100EA 65 USD
TS18023-0100 TS18023-0500
Silver cyanide 99% for analysis
Silver cyanide
Formula: AgCN
MW: 133.89
Melting Pt: >300 °C
Density: 3.943
Storage Temperature: Ambient
MDL Number: MFCD00003409
CAS Number: 506-64-9
EINECS: 208-048-6
UN: 1684
ADR: 6.1 II
Merck Index: 12,08655

Order Now

Specification Test Results

Appearance White Powder
Assay ≥98.5 %
Aluminium (Al) ≤10 ppm
Copper (Cu) ≤10 ppm
Iron (Fe) ≤30 ppm
Lead (Pb) ≤10 ppm
Sodium (Na) ≤500 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR