p-Toluenesulfonic acid monohydrate 97.5%, pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-Methylbenzenesulphonic acid hydrate, Toluene-4-sulphonic acid monohydrate

17178-0010 17178-0025 17178-0050 17178-5000
TS17178-0010EA 57 USD
TS17178-0010 TS17178-0025 TS17178-0050 TS17178-5000
p-Toluenesulfonic acid monohydrate 97.5%, pure
p-Toluenesulphonic acid monohydrate
Formula: C₇H₈O₃S·1H₂O
MW: 190.22 g/mol
Melting Pt: 100…108 °C
Flash Pt: 180 °C
Storage Temperature: Ambient
MDL Number: MFCD00142137
CAS Number: 6192-52-5
EINECS: 203-180-0
UN: 2585
ADR: 8 III
Merck Index: 12,09671

Order Now

Specification Test Results

Appearance White to off-white Adhering crystals (May discolor to pink on storage)
Infrared spectrum Conforms
Titration with NaOH ≥96.5 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR