Dibenzo-18-crown-6-(dibenzo-1,4,7,10,13,16-hexaoxacyclooctadecane, tin(II) ionophore I) 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Dibenzo-18-crown-6

16980-0100 16980-0500
TS16980-0100EA 54 USD
TS16980-0100 TS16980-0500
Dibenzo-18-crown-6-(dibenzo-1,4,7,10,13,16-hexaoxacyclooctadecane, tin(II) ionophore I) 98+%
Dibenzo-18-crown-6-(dibenzo-1,4,7,10,13,16-hexaoxacyclooctadecane, tin(II) ionophore I)
Formula: C₂₀H₂₄O₆
MW: 360.41 g/mol
Boiling Pt: 380…384 °C (679 mmHg)
Melting Pt: 160…164 °C
Storage Temperature: Ambient
MDL Number: MFCD00005098
CAS Number: 14187-32-7
EINECS: 238-041-3

Order Now

Specification Test Results

Appearance White to light beige Powder
Infrared spectrum Conforms
Melting point 160 to 164 °C
HPLC ≥98.0 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR