Deuterium oxide 100.0 atom % D NMR

Supplier: Thermo Scientific Chemicals

Synonyms: Water-D2, Heavy water

16631-1000 16631-0100 166310101 166310500
TS16631-1000EA 408 USD
TS16631-1000 TS16631-0100 TS166310101 TS166310500
Deuterium oxide 100.0 atom % D NMR
Deuterium oxide
Formula: D₂O
MW: 20 g/mol
Boiling Pt: 101.3
Melting Pt: 3.8
Density: 1.105 g/cm³
Storage Temperature: Refrigerator
MDL Number: MFCD00044636
CAS Number: 7789-20-0
EINECS: 232-148-9
Merck Index: 12,02984

Order Now

Specification Test Results

Appearance Clear colorless Liquid
D-Enrichment ≥99.95 %
Melting point 150 to 156 °C
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR