Diethyl mesoxalate (diethyl ketomalonate) 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Diethyl 2-oxomalonate

11443-0050 11443-0250
TS11443-0050EA 48 USD
TS11443-0050 TS11443-0250
Diethyl mesoxalate (diethyl ketomalonate) 95%
Diethyl mesoxalate (diethyl ketomalonate)
Formula: C₇H₁₀O₅
MW: 174.15 g/mol
Boiling Pt: 208…210 °C (760 mmHg)
Density: 1.142 g/cm³
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00009121
CAS Number: 609-09-6
EINECS: 210-176-2

Order Now

Specification Test Results

Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4150 to 1.4190 (20 °C, 589 nm)
GC ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR