Dibenzosuberenone 97%

Supplier: Thermo Scientific Chemicals

Synonyms: 5H-Dibenzo[a,d]cyclohepten-5-one, 5H-Dibenzo[a,d][7]annulen-5-one

11253-1000
TS11253-1000EA 189 USD
TS11253-1000
Dibenzosuberenone 97%
Dibenzosuberenone
Formula: C₁₅H₁₀O
MW: 206.24 g/mol
Boiling Pt: 347 °C (1013 hPa)
Melting Pt: 85…91 °C
MDL Number: MFCD00003588
CAS Number: 2222-33-5
EINECS: 233-774-5

Order Now

Specification Test Results

Appearance White to yellow to beige Crystalline powder
Infrared spectrum Conforms
Melting point 85 to 91 °C
Loss on drying ≤0.5 % (60 °C) (vacuum)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR