Ethylenetetracarbonitrile ≥98.0% (by total nitrogen basis), purified by sublimation

Supplier: TCI America
Danger

Synonyms: Tetracyanoethylene

T3264-1G T3264-5G
TCT3264-1GEA 93.1 USD
TCT3264-1G TCT3264-5G
Ethylenetetracarbonitrile ≥98.0% (by total nitrogen basis), purified by sublimation
Ethylenetetracarbonitrile
Formula: C₆N₄
MW: 128.09 g/mol
Boiling Pt: 223
MDL Number: MFCD00001850
CAS Number: 670-54-2
EINECS: 211-578-0
UN: 3439
ADR: 6.1 I
Merck Index: 14,09195

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Melting point 198.0 to 202.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR