3-(Trifluoromethyl)phenyl isothiocyanate ≥98.0% (by GC)

Supplier: TCI America

Synonyms: α,α,α-Trifluoro-m-tolyl isothiocyanate

T2460-5G
TCT2460-5GEA 40 USD
TCT2460-5G
3-(Trifluoromethyl)phenyl isothiocyanate ≥98.0% (by GC)
3-(Trifluoromethyl)phenyl isothiocyanate

CAS Number: 1840-19-3
MDL Number: MFCD00004806
Molecular Formula: C8H4F3NS
Molecular Weight: 203.18
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Very Pale Yellow
Boiling point (°C): 208
Flash Point (°C): 95
Specific Gravity (20/20): 1.34


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: Isothiocyanic Acid 3-(Trifluoromethyl)phenyl Ester, alpha,alpha,alpha-Trifluoro-m-tolyl Isothiocyanate

Formula: C₈H₄F₃NS
MW: 203.19 g/mol
Boiling Pt: 208 °C
Density: 1.34 (20 °C)
Flash Pt: 95 °C
MDL Number: MFCD00004806
CAS Number: 1840-19-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7c2da9980dd8d912d39f6640f9ee97db434ac976&vid=7c2da9980dd8d912d39f6640f9ee97db434ac976&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR