2-(3-Trifluoromethylanilino)nicotinic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 2-[(3-Trifluoromethylphenyl)amino]nicotinic Acid, 2-((3-(Trifluoromethyl)phenyl)amino)nicotinic acid, Niflumic Acid

T2353-25G
TCT2353-25GEA 121 USD
TCT2353-25G
2-(3-Trifluoromethylanilino)nicotinic acid ≥98.0% (by HPLC, titration analysis)
2-(3-Trifluoromethylanilino)nicotinic acid

CAS Number: 4394-00-7
MDL Number: MFCD00010569
Molecular Formula: C13H9F3N2O2
Molecular Weight: 282.22
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: Slightly Pale Yellow
Melting point (°C): 204


Product Categories: Bioscience, Immune System, Inflammation, Allergy Research Reagents, Inhibitors (for Pharmacological Research), Anti-rheumatoid Arthritis Ingredients, Anti-inflammatory Ingredients
Other Product Names / Synonyms: 2-[(3-Trifluoromethylphenyl)amino]nicotinic Acid, Niflumic Acid

Formula: C₁₃H₉F₃N₂O₂
MW: 282.22 g/mol
Melting Pt: 204 °C
MDL Number: MFCD00010569
CAS Number: 4394-00-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e4c2d5128e2f01fc12f997cd737d110a8f1aa660&vid=e4c2d5128e2f01fc12f997cd737d110a8f1aa660&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR