Isopropyl trifluoroacetate ≥98.0% (by GC)

Supplier: TCI America

Synonyms: Isopropyltrifluoroacetate, Trifluoroacetic isopropyl ester

T1761-25G
TCT1761-25GEA 53 USD
TCT1761-25G
Isopropyl trifluoroacetate ≥98.0% (by GC)
Isopropyl trifluoroacetate

CAS Number: 400-38-4
MDL Number: MFCD00013566
Molecular Formula: C5H7F3O2
Molecular Weight: 156.10
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 73
Flash Point (°C): -9
Specific Gravity (20/20): 1.12


Other Product Names / Synonyms: Trifluoroacetic Acid Isopropyl Ester

Formula: C₅H₇F₃O₂
MW: 156.1
Boiling Pt: 73 °C
Density: 1.12 (20 °C)
Flash Pt: –9 °C
MDL Number: MFCD00013566
CAS Number: 400-38-4
UN: 2924
ADR: 3 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a20376d59cebaef9f37778c510cb0a98fe11af0c&vid=a20376d59cebaef9f37778c510cb0a98fe11af0c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR