Trichloromethiazide ≥99.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 6-Chloro-3-(dichloromethyl)-3,4-dihydro-2H-benzo[e][1,2,4]thiadiazine-7-sulphonamide 1,1-dioxide, 6-Chloro-3-dichloromethyl-3,4-dihydro-2H-1,2,4-benzothiadiazine-7-sulphonamide 1,1-Dioxide

T1318-10G
TCT1318-010GEA 192.5 USD
TCT1318-010G
Trichloromethiazide ≥99.0% (by titrimetric analysis)
Trichloromethiazide

CAS Number: 133-67-5
MDL Number: MFCD00057315
Molecular Formula: C8H8Cl3N3O4S2
Molecular Weight: 380.64
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Color: White


Product Categories: Bioscience, Other Pharmaceutical Ingredients
Other Product Names / Synonyms: 6-Chloro-3-dichloromethyl-3,4-dihydro-2H-1,2,4-benzothiadiazine-7-sulfonamide 1,1-Dioxide

Formula: C₈H₈Cl₃N₃O₄S₂
MW: 380.64 g/mol
MDL Number: MFCD00057315
CAS Number: 133-67-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d335a2c7ecb959efaf94c8bc0228087c1d0e775e&vid=d335a2c7ecb959efaf94c8bc0228087c1d0e775e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR