N-(Trimethylsilyl)morpholine ≥97.0%

Supplier: TCI America

Synonyms: 4-(Trimethylsilyl)morpholine

T1277-5ML T1277-25ML
TCT1277-005MLEA 58.6 USD
TCT1277-005ML TCT1277-025ML
N-(Trimethylsilyl)morpholine ≥97.0%
N-(Trimethylsilyl)morpholine

CAS Number: 13368-42-8 MDL Number: MFCD00010098 Molecular Formula: C7H17NOSi Molecular Weight: 159.30 Purity/Analysis Method: 97.0% (GC) Form: Clear Liquid Color: Colorless Boiling point (°C): 102 Flash Point (°C): 29 Specific Gravity (20/20): 0.90

Formula: C₇H₁₇NOSi
MW: 159.30 g/mol
Boiling Pt: 102 °C
Density: 0.90 (20 °C)
Flash Pt: 29 °C
MDL Number: MFCD00010098
CAS Number: 13368-42-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ae8c6fc35c0448e5a87c817782a2a96bfaaed2f&vid=4ae8c6fc35c0448e5a87c817782a2a96bfaaed2f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR