(Trimethylsilyl)diazomethane ∼10% ∼0.6 mol/L in hexane

Supplier: TCI America

Synonyms: Trimethylsilyldiazomethane, TMS-Diazomethane

T1146-10ML T1146-100ML T1146-25ML
TCT1146-010MLEA 114.12 USD
TCT1146-010ML TCT1146-100ML TCT1146-25ML
(Trimethylsilyl)diazomethane ∼10% ∼0.6 mol/L in hexane
(Trimethylsilyl)diazomethane

CAS Number: 18107-18-1
MDL Number: MFCD00053946
Molecular Formula: C4H10N2Si
Molecular Weight: 114.22
Form: Clear Liquid
Color: Yellow
Flash Point (°C): -20
Storage Temperature: 0-10°C


Other Product Names / Synonyms: TMS-Diazomethane

Formula: C₄H₁₀N₂Si
MW: 114.22 g/mol
Flash Pt: –20 °C
MDL Number: MFCD00053946
CAS Number: 18107-18-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cab81309b6472e72fcc16415885d59863a05fff8&vid=4ed6393819d3a492519bb168c9f6d46577eeca18&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR