Sertraline hydrochloride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: (1S,4S)-1-(3,4-Dichlorophenyl)-1,2,3,4-tetrahydro-4-(methylamino)naphthalene Hydrochloride

S0507-1G
TCS0507-1GEA 64.16 USD
TCS0507-1G
Sertraline hydrochloride ≥98.0% (by HPLC, titration analysis)
Sertraline hydrochloride

CAS Number: 79559-97-0
MDL Number: MFCD00895772
Molecular Formula: C17H17Cl2N
Molecular Weight: 342.69
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 245
Specific rotation [a]20/D: 38 deg (C=2, MeOH)


Product Categories: Bioscience, Neurotransmitter Agent Ingredients
Other Product Names / Synonyms: (1S,4S)-1-(3,4-Dichlorophenyl)-1,2,3,4-tetrahydro-4-(methylamino)naphthalene Hydrochloride

Formula: C₁₇H₁₇Cl₂N
MW: 342.69 g/mol
Melting Pt: 245 °C
MDL Number: MFCD00895772
CAS Number: 79559-97-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f74d5b7ad22e8acb5ef53586ab368dd8e2c4592a&vid=f74d5b7ad22e8acb5ef53586ab368dd8e2c4592a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR