Sulfanilamide ≥99.9% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 4-Aminobenzenesulphonamide

S0381-25G
TCS0381-025GEA 149 USD
TCS0381-025G
Sulfanilamide ≥99.9% (by titrimetric analysis)
Sulphanilamide

[for Diazotization Titration]
CAS Number: 63-74-1
MDL Number: MFCD00007939
Molecular Formula: C6H8N2O2S
Molecular Weight: 172.20
Purity/Analysis Method: >99.9% (T)
Form: Crystal
Color: White


Other Product Names / Synonyms: 4-Aminobenzenesulfonamide

Formula: H₂NC₆H₄SO₂NH₂
MW: 172.21 g/mol
Boiling Pt: 401 °C (1013 hPa)
Melting Pt: 163…167 °C
Density: 1.46 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007939
CAS Number: 63-74-1
EINECS: 200-563-4
Merck Index: 13,09010
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a76bb9ab6bdf23e9d846b24d4f57047dea1d5071&vid=f2f1c5e47a127cd33d3b4a2a31b1057c375456eb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.9% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR