1-Dodecene ≥99.5%

Supplier: TCI America

Synonyms: Dodec-1-ene

S0342-5ML
TCS0342-005MLEA 49 USD
TCS0342-005ML
1-Dodecene ≥99.5%
1-Dodecene

[Standard Material for GC]
CAS Number: 112-41-4
MDL Number: MFCD00008961
Molecular Formula: C12H24
Molecular Weight: 168.32
Purity/Analysis Method: >99.5% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 213
Flash Point (°C): 76
Specific Gravity (20/20): 0.76

Formula: C₁₂H₂₄
MW: 168.32 g/mol
Boiling Pt: 213 °C
Melting Pt: -35 °C
Density: 0.76 (20 °C)
Flash Pt: 76 °C
Storage Temperature: Ambient
MDL Number: MFCD00008961
CAS Number: 112-41-4
EINECS: 203-968-4

Order Now

Specification Test Results

Purity/Analysis Method >99.5% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR