Dodecahydrofluorene (mixture of isomers) ≥97.0% (by GC)

Supplier: TCI America

Synonyms: Perhydrofluorene

P2393-1G
TCP2393-1GEA 96 USD
TCP2393-1G
Dodecahydrofluorene (mixture of isomers) ≥97.0% (by GC)
Dodecahydrofluorene (mixture of isomers)
Formula: C₁₃H₂₂
MW: 178.32 g/mol
Boiling Pt: 253°C
Melting Pt: 15°C
Density: 0.95
Flash Pt: 64°C
MDL Number: MFCD00001161
CAS Number: 5744-03-6
EINECS: 227-268-3

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 % (total of isomer)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR