4-(4-Piperidyl)pyridine ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 4-(4-Pyridyl)piperidine, 4-(Piperidin-4-yl)pyridine

P2012-5G
TCP2012-5GEA 623.55 USD
TCP2012-5G
4-(4-Piperidyl)pyridine ≥98.0% (by titrimetric analysis)
4-(4-Piperidyl)pyridine

CAS Number: 581-45-3
MDL Number: MFCD05863877
Molecular Formula: C10H14N2
Molecular Weight: 162.24
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Boiling point (°C): 292
Melting point (°C): 86


Other Product Names / Synonyms: 4-(4-Pyridyl)piperidine

Formula: C₁₀H₁₄N₂
MW: 162.24 g/mol
Boiling Pt: 292 °C
Melting Pt: 86 °C
MDL Number: MFCD05863877
CAS Number: 581-45-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=afdbd33f6d41c045b4ad9d06b41c2799eba6b6f1&vid=afdbd33f6d41c045b4ad9d06b41c2799eba6b6f1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR