Pivalamidine hydrochloride ≥98.0%

Supplier: TCI America

Synonyms: Pivalimidamide hydrochloride, 2,2-Dimethylpropionamidine Hydrochloride

P1761-25G P1761-5G
TCP1761-25GEA 243.8 USD
TCP1761-25G TCP1761-5G
Pivalamidine hydrochloride ≥98.0%
Pivalamidine hydrochloride

CAS Number: 18202-73-8
MDL Number: MFCD00051988
Molecular Formula: C5H12N2
Molecular Weight: 136.62
Purity/Analysis Method: >98.0% (N,T)
Form: Crystal
Color: White
Melting point (°C): 193


Other Product Names / Synonyms: 2,2-Dimethylpropionamidine Hydrochloride

Formula: C₅H₁₂N₂
MW: 136.62 g/mol
Melting Pt: 193 °C
MDL Number: MFCD00051988
CAS Number: 18202-73-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9947c683c3c1685af3cd1607bd2ff458936228ca&vid=9947c683c3c1685af3cd1607bd2ff458936228ca&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (N,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR