POPSO (β,β'-Dihydroxypiperazine-1,4-dipropanesulfonic acid)

Supplier: TCI America

Synonyms: 3,3'-(Piperazine-1,4-diyl)bis(2-hydroxypropane-1-sulphonic acid), Piperazine-1,4-bis(2-hydroxypropanesulphonic acid) dihydrate, POPSO

P1131-25G
TCP1131-025GEA 58.17 USD
TCP1131-025G
POPSO (β,β'-Dihydroxypiperazine-1,4-dipropanesulfonic acid)
POPSO (β,β'-dihydroxypiperazine-1,4-dipropanesulphonic acid)

[Good's buffer component for biological research]
CAS Number: 68189-43-5
MDL Number: MFCD00038350
Molecular Formula: C10H22N2O8S2
Molecular Weight: 362.41
Form: Crystal
Color: White


Other Product Names / Synonyms: POPSO

Formula: C₁₀H₂₂N₂O₈S₂
MW: 362.4203 g/mol
MDL Number: MFCD00038350
CAS Number: 68189-43-5
EINECS: 269-199-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=376734e754e64e39c76253c599e84cf2f852ae2c&vid=d79bd2d7dfd1c3de9c9339a77a4b01c215545dbf&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR