Monoethyl-4-oxoheptanedioate ≥97.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 7-Ethoxy-4,7-dioxoheptanoic acid, 4-Oxoheptanedioic Acid Monoethyl Ester

O0211-5G
TCO0211-005GEA 285.6 USD
TCO0211-005G
Monoethyl-4-oxoheptanedioate ≥97.0% (by titrimetric analysis)
Monoethyl-4-oxoheptanedioate

CAS Number: 1506-55-4
MDL Number: MFCD00059093
Molecular Formula: C9H14O5
Molecular Weight: 202.21
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 70


Other Product Names / Synonyms: 4-Oxoheptanedioic Acid Monoethyl Ester

Formula: C₉H₁₄O₅
MW: 202.21 g/mol
Melting Pt: 70 °C
MDL Number: MFCD00059093
CAS Number: 1506-55-4

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR