4-(1-Naphthyl)-1-naphthaleneboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

N1010-1G
TCN1010-1GEA 295.61 USD
TCN1010-1G
4-(1-Naphthyl)-1-naphthaleneboronic acid (contains varying amounts of Anhydride)
4-(1-Naphthyl)-1-naphthaleneboronic acid
Formula: C₂₀H₁₅BO₂
MW: 298.15 g/mol
MDL Number: MFCD16294426
CAS Number: 363607-69-6
New Product

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 107.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR