(4-Nitrophenyl)acetic acid ethyl ester ≥98.0%

Supplier: TCI America

Synonyms: Ethyl 4-nitrohenylacetate, Ethyl (4-nitrophenyl)acetate, Ethyl 4-nitrophenylacetate

N0228-25G N0228-5G
TCN0228-025GEA 37 USD
TCN0228-025G TCN0228-5G
(4-Nitrophenyl)acetic acid ethyl ester ≥98.0%
(4-Nitrophenyl)acetic acid ethyl ester

CAS Number: 5445-26-1
MDL Number: MFCD00017046
Molecular Formula: C10H11NO4
Molecular Weight: 209.20
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 196
Melting point (°C): 64


Other Product Names / Synonyms: 4-Nitrophenylacetic Acid Ethyl Ester

Formula: C₁₀H₁₁NO₄
MW: 209.2 g/mol
Boiling Pt: 196 °C
Melting Pt: 64 °C
MDL Number: MFCD00017046
CAS Number: 5445-26-1
EINECS: 226-646-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a1e58262c8eb2f281e3a4188c465582e0c3e3c9d&vid=a1e58262c8eb2f281e3a4188c465582e0c3e3c9d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR