Maleamide

Supplier: TCI America

Synonyms: Maleic Acid Diamide

M0004-25G
TCM0004-25GEA 152.7 USD
TCM0004-25G
Maleamide
Maleamide

CAS Number: 928-01-8
MDL Number: MFCD00082355
Molecular Formula: C4H6N2O2
Molecular Weight: 114.10
Form: Crystal
Color: White


Other Product Names / Synonyms: Maleic Acid Diamide

Formula: C₄H₆N₂O₂
MW: 114.10 g/mol
MDL Number: MFCD00082355
CAS Number: 928-01-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2939085166cd2e2d34ecdf11b1021a5569ad76c9&vid=2939085166cd2e2d34ecdf11b1021a5569ad76c9&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR