Calcium-3-methyl-2-oxobutyrate hydrate ≥97.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 3-Methyl-2-oxobutyric Acid Calcium Salt

K0021-5G
TCK0021-005GEA 141.2 USD
TCK0021-005G
Calcium-3-methyl-2-oxobutyrate hydrate ≥97.0% (by titrimetric analysis)
Calcium-3-methyl-2-oxobutyrate hydrate

MDL Number: MFCD00002580
Molecular Formula: C5H8O3
Molecular Weight: 270.29
Purity/Analysis Method: >97.0% (T)
Form: Crystal


Other Product Names / Synonyms: 3-Methyl-2-oxobutyric Acid Calcium Salt

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=08488412b949a6092a34769a5e8acb83a82adf0f&vid=ac310f7506e060a24aac6840b74c17a065d93f0b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR