Sodium dodecyl sulphate (SDS) 97,0

Supplier: TCI America
Danger

Synonyms: Dodecyl sulphate sodium salt, Sodium dodecyl sulfate, Dodecyl sulphuric acid sodium salt, Lauryl sulphate sodium salt, Sodium lauryl sulphate

I0352-500G I0352-25G I0352-100G
TCI0352-500GEA 166.7 USD
TCI0352-500G TCI0352-025G TCI0352-100G
Sodium dodecyl sulphate (SDS) 97,0
Sodium dodecyl sulphate (SDS)

[Reagent for Ion-Pair Chromatography]
CAS Number: 151-21-3
MDL Number: MFCD00036175
Molecular Formula: C12H26O4S
Molecular Weight: 288.38
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 207
Flash Point (°C): 145


Product Categories: Analytical Science, HPLC Ion-Pair Reagents for Basic Samples
Other Product Names / Synonyms: IPC-SDS, Laurylsulfuric Acid Sodium Salt, Dodecylsulfuric Acid Sodium Salt, Sodium Lauryl Sulfate

Formula: CH₃(CH₂)₁₁OSO₃Na
MW: 288.38 g/mol
Melting Pt: 207 °C
Density: 1.1 g/cm³ (20 °C)
Flash Pt: 145 °C
Storage Temperature: Ambient
MDL Number: MFCD00036175
CAS Number: 151-21-3
EINECS: 205-788-1
UN: 1325
ADR: 4.1 II
Merck Index: 14,08636
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=70133d1a576563b95828e0ed37231b08216ed2af&vid=0596d7502d61df67d5e9e3d5cf2392e080493266&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR