(±)-3-Hexanol ≥98.0% (by GC)

Supplier: TCI America

Synonyms: (±)-Propyl ethyl carbinol, (±)-Ethyl propyl carbinol, (±)-3-Hydroxy hexane

H0132-25ML
TCH0132-25MLEA 59.6 USD
TCH0132-25ML
(±)-3-Hexanol ≥98.0% (by GC)
(±)-3-Hexanol

CAS Number: 623-37-0
MDL Number: MFCD00004582
Molecular Formula: C6H14O
Molecular Weight: 102.18
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 135
Flash Point (°C): 45
Specific Gravity (20/20): 0.82


Other Product Names / Synonyms: 3-Hexyl Alcohol

Formula: C₆H₁₄O
MW: 102.18 g/mol
Boiling Pt: 135 °C
Melting Pt: -70
Density: 0.82 (20 °C)
Flash Pt: 45 °C
MDL Number: MFCD00004582
CAS Number: 623-37-0
UN: 2282
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=39530c9bef58d2cdcee4bbc14b525d77fdf89c21&vid=f007f095722cbdace163e1263edb2bf7a2739678&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR