Magnesium(II) gluconate hydrate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: D-Gluconic acid hemimagnesium salt hydrate, Magnesium D-gluconate hydrate, Gluconic acid magnesium(II) salt hydrate, D-Gluconic acid magnesium salt hydrate

G0276-25G G0276-500G
TCG0276-25GEA 48 USD
TCG0276-25G TCG0276-500G
Magnesium(II) gluconate hydrate ≥98.0% (by titrimetric analysis)
Magnesium(II) gluconate hydrate

CAS Number: 3632-91-5
MDL Number: MFCD00150971
Molecular Formula: C6H12O7
Molecular Weight: 414.60
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Specific rotation [a]20/D: 12 deg (C=1, H2O)


Product Categories: Bioscience, Sugar Acids, Glucose
Other Product Names / Synonyms: Gluconic Acid Magnesium(II) Salt

Formula: C₆H₁₂O₇·xH₂O
MW: 414.60 g/mol (anhy)
MDL Number: MFCD00150971
CAS Number: 3632-91-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9cf5b3aecae7f7f9115340d091c64eaafeff8e52&vid=7a7b99536185e3d7234079074d07461eb970985c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR