Glycyl-4-aminobutyric acid

Supplier: TCI America

G0115-1G
TCG0115-001GEA 76 USD
TCG0115-001G
Glycyl-4-aminobutyric acid
Glycyl-4-aminobutyric acid

CAS Number: 32595-49-6 MDL Number: MFCD00056123 Molecular Formula: C6H12N2O3 Molecular Weight: 160.17 Form: Crystal Color: White

Formula: C₆H₁₂N₂O₃
MW: 160.17 g/mol
MDL Number: MFCD00056123
CAS Number: 32595-49-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=15d2dac575c598c3c01ba5b3f3d943c76c2499f6&vid=15d2dac575c598c3c01ba5b3f3d943c76c2499f6&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR