Iron(II) gluconate hydrate ≥95.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Iron(II) Gluconate Hydrate, Iron(II) D-gluconate dihydrate, Gluconic Acid Iron(II) Salt

G0038-500G G0038-25G
TCG0038-500GEA 146 USD
TCG0038-500G TCG0038-25G
Iron(II) gluconate hydrate ≥95.0% (by titrimetric analysis)
Iron(II) gluconate hydrate

CAS Number: 22830-45-1
MDL Number: MFCD00150872
Molecular Formula: C6H12O7
Molecular Weight: 446.14
Purity/Analysis Method: >95.0% (T)
Form: Crystal


Product Categories: Bioscience, Sugar Acids, Glucose
Other Product Names / Synonyms: Gluconic Acid Iron(II) Salt

Formula: C₁₂H₂₄FeO₁₅
MW: 464.16 g/mol
MDL Number: MFCD00150872
CAS Number: 22830-45-1

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR