7-Fluoroindole ≥98.0%

Supplier: TCI America

F0555-1G
TCF0555-1GEA 141.2 USD
TCF0555-1G
7-Fluoroindole ≥98.0%
7-Fluoroindole

CAS Number: 387-44-0
MDL Number: MFCD01074502
Molecular Formula: C8H6FN
Molecular Weight: 135.14
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 67

Formula: C₈H₆FN
MW: 135.14 g/mol
Melting Pt: 67 °C
MDL Number: MFCD01074502
CAS Number: 387-44-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ef985e151021e48ce805d6ff4abfc15a6beb1076&vid=ef985e151021e48ce805d6ff4abfc15a6beb1076&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR