Ethylene sulfide ≥98.0% stabilized

Supplier: TCI America

Synonyms: Dimethylene Sulphide, Thiirane

E0133-10G
TCE0133-010GEA 141.74 USD
TCE0133-010G
Ethylene sulfide ≥98.0% stabilized
Ethylene sulfide

(stabilized with Butyl Mercaptan)
CAS Number: 420-12-2
MDL Number: MFCD00005159
Molecular Formula: C2H4S
Molecular Weight: 60.11
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 56
Flash Point (°C): 10
Freezing point (°C): -109
Specific Gravity (20/20): 1.01
Storage Temperature: <0°C


Other Product Names / Synonyms: Dimethylene Sulfide, Thiirane

Formula: C₂H₄S
MW: 60.11 g/mol
Boiling Pt: 56 °C
Density: 1.01 (20 °C)
Flash Pt: 10 °C
MDL Number: MFCD00005159
CAS Number: 420-12-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f152ff48da3c53b86eb00ca3c21db47f2aa725af&vid=f152ff48da3c53b86eb00ca3c21db47f2aa725af&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR