4-(Dimethylamino)butyraldehyde diethyl acetal ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 4,4-Diethoxy-N,N-dimethylbutylamine, 4,4-Diethoxy-N,N-dimethyl-1-butanamine

D3973-5G
TCD3973-5GEA 33 USD
TCD3973-5G
4-(Dimethylamino)butyraldehyde diethyl acetal ≥98.0% (by GC, titration analysis)
4-(Dimethylamino)butyraldehyde diethyl acetal

CAS Number: 1116-77-4
MDL Number: MFCD00671479
Molecular Formula: C10H23NO2
Molecular Weight: 189.30
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 95
Flash Point (°C): 70
Specific Gravity (20/20): 0.86
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 4,4-Diethoxy-N,N-dimethylbutylamine

Formula: C₁₀H₂₃NO₂
MW: 189.30 g/mol
Boiling Pt: 95 °C
Density: 0.86 (20 °C)
Flash Pt: 70 °C
MDL Number: MFCD00671479
CAS Number: 1116-77-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=57e42b611cd06e905f1a754e1fbb029d6741f317&vid=57e42b611cd06e905f1a754e1fbb029d6741f317&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR