3,5-Dibromoanisole ≥98.0%

Supplier: TCI America

Synonyms: 1,3-Dibromo-5-methoxybenzene

D3364-5G
TCD3364-5GEA 45 USD
TCD3364-5G
3,5-Dibromoanisole ≥98.0%
3,5-Dibromoanisole

CAS Number: 74137-36-3
MDL Number: MFCD02258848
Molecular Formula: C7H6Br2O
Molecular Weight: 265.93
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 261
Melting point (°C): 39
Flash Point (°C): 110


Other Product Names / Synonyms: 1,3-Dibromo-5-methoxybenzene

Formula: C₇H₆Br₂O
MW: 265.93 g/mol
Boiling Pt: 261 °C
Melting Pt: 39 °C
Flash Pt: 110 °C
MDL Number: MFCD02258848
CAS Number: 74137-36-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=27e1963e65434cdf217175032fd2c7619a8cb7c3&vid=27e1963e65434cdf217175032fd2c7619a8cb7c3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR