Dibenzo-21-crown-7-(dibenzo-1,4,7,10,13,16,19-heptaoxacycloheneicosane) 96,0

Supplier: TCI America

Synonyms: 6,7,9,10,12,13,20,21,23,24-Decahydrodibenzo[b,k][1,4,7,10,13,16,19]heptaoxacyclohenicosine

D2878-1G
TCD2878-1GEA 223 USD
TCD2878-1G
Dibenzo-21-crown-7-(dibenzo-1,4,7,10,13,16,19-heptaoxacycloheneicosane) 96,0
Dibenzo-21-crown-7-(dibenzo-1,4,7,10,13,16,19-heptaoxacycloheneicosane)

CAS Number: 14098-41-0
MDL Number: MFCD00068646
Molecular Formula: C22H28O7
Molecular Weight: 404.46
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Melting point (°C): 107

Formula: C₂₂H₂₈O₇
MW: 404.46 g/mol
Melting Pt: 107 °C
MDL Number: MFCD00068646
CAS Number: 14098-41-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=28f1c6d38eff6417bdd049aa014fc3c4b384ddf0&vid=28f1c6d38eff6417bdd049aa014fc3c4b384ddf0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR