2,4-Dimethoxycinnamic acid ≥96.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 3-(2,4-Dimethoxyphenyl)acrylic acid

D2364-25G
TCD2364-025GEA 174.7 USD
TCD2364-025G
2,4-Dimethoxycinnamic acid ≥96.0% (by GC, titration analysis)
2,4-Dimethoxycinnamic acid

CAS Number: 6972-61-8
MDL Number: MFCD00016837
Molecular Formula: C11H12O4
Molecular Weight: 208.21
Purity/Analysis Method: >96.0% (GC,T)
Form: Crystal
Melting point (°C): 184

Formula: C₁₁H₁₂O₄
MW: 208.21 g/mol
Melting Pt: 184 °C
MDL Number: MFCD00016837
CAS Number: 16909-09-4
EINECS: 240-955-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c749714d0761909fbf2767c381c1d10eafb91db4&vid=2fc7f505f045ca7700226bd35e3c69db17098f5d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >96.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR