4,4'-Dibromooctafluorobiphenyl (DBOB) ≥95.0% (by GC)

Supplier: TCI America

Synonyms: 4,4-Dibromooctafluorobiphenyl

D1696-1G
TCD1696-1GEA 39 USD
TCD1696-1G
4,4'-Dibromooctafluorobiphenyl (DBOB) ≥95.0% (by GC)
4,4'-Dibromooctafluorobiphenyl (DBOB)

CAS Number: 10386-84-2
MDL Number: MFCD00000310
Molecular Formula: C12Br2F8
Molecular Weight: 455.93
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Melting point (°C): 115

Formula: C₁₂Br₂F₈
MW: 455.94 g/mol
Melting Pt: 115 °C
MDL Number: MFCD00000310
CAS Number: 10386-84-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c210eaa46cf493960e29848bfca87724927fec2e&vid=c210eaa46cf493960e29848bfca87724927fec2e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR